4-Chloro-3-(trifluoromethyl)phenol 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1930L-100G
PC1930L-25G
PC1930L-5G
APOSPC1930L-100GEA
2400
CZK
APOSPC1930L-100G
APOSPC1930L-25G
APOSPC1930L-5G
4-Chloro-3-(trifluoromethyl)phenol 98%
4-Chloro-3-(trifluoromethyl)phenol
Vzorec:
C₇H₄ClF₃O Bod varu: 120…122 °C/27mmHg Bod tání: 42…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042525 CAS číslo: 6294-93-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...