Tisk…

2-Chlor-4-fluorothiophenol 97%

Dodavatel: Apollo Scientific
Varování

PC1889-1G PC1889-250MG PC1889-5G
APOSPC1889-1GEA 1740 CZK
APOSPC1889-1G APOSPC1889-250MG APOSPC1889-5G
2-Chlor-4-fluorothiophenol 97%
2-Chlor-4-fluorothiophenol
Vzorec: C₆H₄ClFS
Bod varu: 86…88 °C/15mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00052936
CAS číslo: 175277-99-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR