3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1886P-1G
PC1886P-5G
PC1886P-500MG
APOSPC1886P-1GEA
2480
CZK
APOSPC1886P-1G
APOSPC1886P-5G
APOSPC1886P-500MG
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride 97%
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride
Vzorec:
C₁₁H₆Cl₂FNO₂ MW: 274,08 g/mol Bod tání: 49…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055650 CAS číslo: 69399-79-7 EINECS: 273-987-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...