Tisk…

3-Chloro-4-fluorophenyl isocyanate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-1-fluoro-4-isocyanatobenzene

PC1886-25G PC1886-5G
APOSPC1886-25GEA 8830 CZK
APOSPC1886-25G APOSPC1886-5G
3-Chloro-4-fluorophenyl isocyanate 98%
3-Chloro-4-fluorophenyl isocyanate

  • May form small amount of sediment.

Vzorec: C₇H₃ClFNO
Bod varu: 60…62 °C/2mmHg
Hustota: 1,377 g/cm³ (25 °C)
Bod vzplanutí: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037037
CAS číslo: 50529-33-4
UN: 2206
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR