Tisk…

3-Chloro-4-fluorophenylhydrazine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

PC1884S-1G PC1884S-5G
APOSPC1884S-1GEA 622 CZK
APOSPC1884S-1G APOSPC1884S-5G
3-Chloro-4-fluorophenylhydrazine hydrochloride 97%
3-Chloro-4-fluorophenylhydrazine hydrochloride
Vzorec: C₆H₇Cl₂FN₂
Storage Temperature: Ambient
MDL Number: MFCD00052267
CAS číslo: 175135-74-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR