Tisk…

4-(2-Chloro-6-fluorophenyl)but-3-en-2-one 97%

Dodavatel: Apollo Scientific

PC1884E-1G PC1884E-5G
APOSPC1884E-1GEA 830 CZK
APOSPC1884E-1G APOSPC1884E-5G
4-(2-Chloro-6-fluorophenyl)but-3-en-2-one 97%
4-(2-Chloro-6-fluorophenyl)but-3-en-2-one
Vzorec: C₁₀H₈ClFO
MW: 198,63 g/mol
Bod varu: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00051608
CAS číslo: 175136-82-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR