Tisk…

2-Hydroxy-6-(trifluoromethyl)pyridine-3-carboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydroxy-6-trifluoromethylnicotinic acid, 2-Hydroxy-6-(trifluoromethyl)nicotinic acid

PC1877-10G PC1877-1G PC1877-5G
APOSPC1877-10GEA 3190 CZK
APOSPC1877-10G APOSPC1877-1G APOSPC1877-5G
2-Hydroxy-6-(trifluoromethyl)pyridine-3-carboxylic acid 98%
2-Hydroxy-6-(trifluoromethyl)pyridine-3-carboxylic acid
Vzorec: C₇H₄F₃NO₃
MW: 207,11 g/mol
Bod tání: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD01862657
CAS číslo: 191595-63-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR