Tisk…

1-Chloro-5-fluoro-2-iodobenzene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-4-fluoro-1-iodobenzene, 2-Chloro-4-fluoroiodobenzene

PC1868H-25G PC1868H-100G PC1868H-500G
APOSPC1868H-25GEA 744 CZK
APOSPC1868H-25G APOSPC1868H-100G APOSPC1868H-500G
1-Chloro-5-fluoro-2-iodobenzene 98%
1-Chloro-5-fluoro-2-iodobenzene
Vzorec: C₆H₃ClFI
MW: 256.45
Bod varu: 96…98 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00051774
CAS číslo: 101335-11-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR