2-Chloro-6-fluorobenzyl mercaptan 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloro-6-fluorobenzylthiol
PC1864N-1G
PC1864N-5G
APOSPC1864N-1GEA
3190
CZK
APOSPC1864N-1G
APOSPC1864N-5G
2-Chloro-6-fluorobenzyl mercaptan 97%
2-Chloro-6-fluorobenzyl mercaptan
Vzorec:
C₇H₆ClFS Bod varu: 102 °C/15mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD00052473 CAS číslo: 170924-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...