α,2-Dichloro-6-fluorotoluene 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-6-fluorobenzyl chloride, 1-Chloro-2-(chloromethyl)-3-fluorobenzene
PC1864H-100G
PC1864H-250G
PC1864H-25G
APOSPC1864H-100GEA
1240
CZK
APOSPC1864H-100G
APOSPC1864H-250G
APOSPC1864H-25G
α,2-Dichloro-6-fluorotoluene 97%
α,2-Dichloro-6-fluorotoluene
Vzorec:
C₇H₅Cl₂F MW: 179,02 g/mol Bod varu: 208…210 °C Hustota: 1,401 g/cm³ (25 °C) Bod vzplanutí: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000896 CAS číslo: 55117-15-2 EINECS: 259-487-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...