3-Chloro-4-fluorobenzylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1864F-1G
PC1864F-25G
PC1864F-5G
APOSPC1864F-1GEA
622
CZK
APOSPC1864F-1G
APOSPC1864F-25G
APOSPC1864F-5G
3-Chloro-4-fluorobenzylamine 97%
3-Chloro-4-fluorobenzylamine
Vzorec:
C₇H₇ClFN MW: 159,59 g/mol Bod varu: 104…106 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00042507 CAS číslo: 72235-56-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...