2-Chloro-4-fluorobenzylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1864B-25G
PC1864B-100G
APOSPC1864B-25GEA
1080
CZK
APOSPC1864B-25G
APOSPC1864B-100G
2-Chloro-4-fluorobenzylamine 97%
2-Chloro-4-fluorobenzylamine
Vzorec:
C₇H₇ClFN MW: 159,59 g/mol Bod varu: 94…96 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00042532 CAS číslo: 15205-11-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...