Tisk…

2-Chloro-4-fluorobenzoyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Chloro-4-fluorobenzene-1-carbonyl chloride, 2-Chloro-4-fluorobenzoylchloride

PC1863T-25G PC1863T-5G PC1863T-100G
APOSPC1863T-25GEA 1240 CZK
APOSPC1863T-25G APOSPC1863T-5G APOSPC1863T-100G
2-Chloro-4-fluorobenzoyl chloride 98%
2-Chloro-4-fluorobenzoyl chloride
Vzorec: C₇H₃Cl₂FO
MW: 193 g/mol
Bod varu: 82…84 °C/15mmHg
Hustota: 1,447 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00084954
CAS číslo: 21900-54-9
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR