2-Chloro-4-fluorobenzonitrile 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1861L-100G
PC1861L-25G
PC1861L-250G
APOSPC1861L-100GEA
4070
CZK
APOSPC1861L-100G
APOSPC1861L-25G
APOSPC1861L-250G
2-Chloro-4-fluorobenzonitrile 95%
2-Chloro-4-fluorobenzonitrile
Vzorec:
C₇H₃ClFN Bod tání: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042523 CAS číslo: 60702-69-4 EINECS: 262-384-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...