Tisk…

1-[(2-Chloroethyl)sulphonyl]-4-fluorobenzene 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Chloroethyl 4-fluorophenyl sulphone

PC1812R-1G PC1812R-5G
APOSPC1812R-1GEA 2280 CZK
APOSPC1812R-1G APOSPC1812R-5G
1-[(2-Chloroethyl)sulphonyl]-4-fluorobenzene 98%
1-[(2-Chloroethyl)sulphonyl]-4-fluorobenzene
Vzorec: C₈H₈ClFO₂S
MW: 222,67 g/mol
Bod tání: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00203483
CAS číslo: 33330-46-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR