4-Chloro-2-(trifluoromethyl)phenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane
PC1777-5G
PC1777-100G
PC1777-25G
APOSPC1777-5GEA
622
CZK
APOSPC1777-5G
APOSPC1777-100G
APOSPC1777-25G
4-Chloro-2-(trifluoromethyl)phenylboronic acid 98%
4-Chloro-2-(trifluoromethyl)phenylboronic acid
Vzorec:
C₇H₅BClF₃O₂ MW: 224.37 Bod tání: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973086 CAS číslo: 313545-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...