Potassium-2-(ethoxycarbonyl)phenyltrifluoroborate 98%
Dodavatel: Apollo Scientific
Synonyms:
Potassium (2-(ethoxycarbonyl)phenyl)trifluoroborate
PC1768-5G
APOSPC1768-5GEA
2240
CZK
APOSPC1768-5G
Potassium-2-(ethoxycarbonyl)phenyltrifluoroborate 98%
Potassium-2-(ethoxycarbonyl)phenyltrifluoroborate
Vzorec:
C₉H₉BF₃KO₂ MW: 256,0711 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115768 CAS číslo: 850623-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...