4-Chloro-2,5-difluorobenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC1757-25G
APOSPC1757-25GEA
2570
CZK
APOSPC1757-25G
4-Chloro-2,5-difluorobenzoic acid 98%
4-Chloro-2,5-difluorobenzoic acid
Vzorec:
C₇H₃ClF₂O₂ MW: 192,55 g/mol Bod tání: 153…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077480 CAS číslo: 132794-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...