Tisk…

Potassium-4-(ethylthio)phenyltrifluoroborate 96%

Dodavatel: Apollo Scientific

PC1733-10G PC1733-25G PC1733-5G
APOSPC1733-10GEA 7480 CZK
APOSPC1733-10G APOSPC1733-25G APOSPC1733-5G
Potassium-4-(ethylthio)phenyltrifluoroborate 96%
Potassium-4-(ethylthio)phenyltrifluoroborate
Vzorec: C₈H₉BF₃KS
Storage Temperature: Ambient
MDL Number: MFCD04115771
CAS číslo: 850623-75-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR