Tisk…

Potassium-2-(aminocarbonyl)phenyltrifluoroborate 96%

Dodavatel: Apollo Scientific

PC1727-1G PC1727-250MG
APOSPC1727-1GEA 8100 CZK
APOSPC1727-1G APOSPC1727-250MG
Potassium-2-(aminocarbonyl)phenyltrifluoroborate 96%
Potassium-2-(aminocarbonyl)phenyltrifluoroborate
Vzorec: C₇H₆BF₃KNO
Storage Temperature: Ambient
MDL Number: MFCD04115765
CAS číslo: 850623-70-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR