3-Chloro-4,5-diaminobenzotrifluoride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Chloro-5-trifluoromethyl-o-phenylenediamine
PC1716-100G
PC1716-25G
APOSPC1716-100GEA
15970
CZK
APOSPC1716-100G
APOSPC1716-25G
3-Chloro-4,5-diaminobenzotrifluoride 98%
3-Chloro-4,5-diaminobenzotrifluoride
Vzorec:
C₇H₆ClF₃N₂ MW: 210,59 g/mol Bod tání: 40…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042198 CAS číslo: 132915-80-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...