2-Chloro-4-(trifluoromethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Chloro-4-cyanobenzotrifluoride
PC1713-100G
PC1713-25G
PC1713-5G
APOSPC1713-100GEA
5430
CZK
APOSPC1713-100G
APOSPC1713-25G
APOSPC1713-5G
2-Chloro-4-(trifluoromethyl)benzonitrile 98%
2-Chloro-4-(trifluoromethyl)benzonitrile
Vzorec:
C₈H₃ClF₃N MW: 205,57 g/mol Bod varu: 192…193 °C Hustota: 1,389 g/cm³ (25 °C) Bod vzplanutí: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084939 CAS číslo: 1813-33-8 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...