Tisk…

Potassium-4-butoxyphenyltrifluoroborate 96%

Dodavatel: Apollo Scientific

PC1707-1G PC1707-5G
APOSPC1707-1GEA 3580 CZK
APOSPC1707-1G APOSPC1707-5G
Potassium-4-butoxyphenyltrifluoroborate 96%
Potassium-4-butoxyphenyltrifluoroborate
Vzorec: C₁₀H₁₃BF₃KO
Bod tání: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115755
CAS číslo: 850623-61-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR