Potassium(2,4-dichlorophenyl)trifluoroborate 96%
Dodavatel: Apollo Scientific
Synonyms:
Potassium (2,4-dichlorophenyl)trifluoroborate, (2,4-Dichlorophenyl)trifluoroboric acid potassium salt
PC1701-1G
PC1701-25G
PC1701-5G
APOSPC1701-1GEA
1490
CZK
APOSPC1701-1G
APOSPC1701-25G
APOSPC1701-5G
Potassium(2,4-dichlorophenyl)trifluoroborate 96%
Potassium(2,4-dichlorophenyl)trifluoroborate
Vzorec:
C₆H₃BCl₂F₃K MW: 252,9 g/mol Bod tání: 98…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115745 CAS číslo: 192863-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...