Tisk…

Cerium trifluoride, anhydrous 99,5%

Dodavatel: Apollo Scientific
Varování

Synonyms: Cerous fluoride, Cerium (III) fluoride

PC1680-250G PC1680-100G PC1680-1KG
APOSPC1680-250GEA 6220 CZK
APOSPC1680-250G APOSPC1680-100G APOSPC1680-1KG
Cerium trifluoride, anhydrous 99,5%
Cerium trifluoride
Vzorec: CeF₃
MW: 197,11 g/mol
Bod varu: 2300 °C
Bod tání: 1640 °C
Hustota: 6,16 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010932
CAS číslo: 7758-88-5
EINECS: 231-841-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR