3-Chloro-4-(trifluoromethyl)phenylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Chloro-4-(trifluoromethyl)benzeneboronic acid
PC1675-1G
PC1675-5G
PC1675-10G
APOSPC1675-1GEA
910
CZK
APOSPC1675-1G
APOSPC1675-5G
APOSPC1675-10G
3-Chloro-4-(trifluoromethyl)phenylboronic acid 97%
3-Chloro-4-(trifluoromethyl)phenylboronic acid
Vzorec:
C₇H₅BClF₃O₂ MW: 224,38 g/mol Bod tání: 206…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788417 CAS číslo: 847756-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...