Tisk…

2-Fluoro-(5-methoxycarbonyl)phenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-5-(methoxycarbonyl)phenylboronic acid, (2-Fluoro-5-(methoxycarbonyl)phenyl)boronic acid

PC1656-1G
APOSPC1656-1GEA 7910 CZK
APOSPC1656-1G
2-Fluoro-(5-methoxycarbonyl)phenylboronic acid 97%
2-Fluoro-(5-methoxycarbonyl)phenylboronic acid
Vzorec: C₈H₈BFO₄
MW: 197,9600 g/mol
Bod tání: 210…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659868
CAS číslo: 850568-04-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR