Tisk…

Calcium trifluoromethanesulphonate 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC1648M-25G PC1648M-100G PC1648M-5G
APOSPC1648M-25GEA 2650 CZK
APOSPC1648M-25G APOSPC1648M-100G APOSPC1648M-5G
Calcium trifluoromethanesulphonate 98%
Calcium trifluoromethanesulphonate
Vzorec: C₂CaF₆O₆S₂
Bod tání: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD00143911
CAS číslo: 55120-75-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR