Tisk…

Calcium tetrafluoroborate 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC1648F-100G PC1648F-25G PC1648F-5G
APOSPC1648F-100GEA 14060 CZK
APOSPC1648F-100G APOSPC1648F-25G APOSPC1648F-5G
Calcium tetrafluoroborate 95%
Calcium tetrafluoroborate
Vzorec: Ca(BF₄)₂
MW: 213,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015972
CAS číslo: 13814-93-2
UN: 3260
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR