Tisk…

(3-Fluoro-4-hydroxyphenyl)boronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Fluoro-4-hydroxyphenylboronic acid, 3-Fluoro-4-hydroxybenzeneboronic acid

PC1628-1G PC1628-5G PC1628-10G
APOSPC1628-1GEA 1290 CZK
APOSPC1628-1G APOSPC1628-5G APOSPC1628-10G
(3-Fluoro-4-hydroxyphenyl)boronic acid 97%
(3-Fluoro-4-hydroxyphenyl)boronic acid
Vzorec: C₅H₅BFNO₂
MW: 155.92 g/mol
Bod tání: 252…256 °C
Storage Temperature: Ambient
MDL Number: MFCD06659838
CAS číslo: 182344-14-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR