3-Fluoro-5-methylphenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Fluoro-5-methylphenyl)boronic acid, 3-Fluoro-5-methylbenzeneboronic acid
PC1610-1G
PC1610-25G
PC1610-5G
APOSPC1610-1GEA
705
CZK
APOSPC1610-1G
APOSPC1610-25G
APOSPC1610-5G
3-Fluoro-5-methylphenylboronic acid 98%
3-Fluoro-5-methylphenylboronic acid
Vzorec:
C₇H₈BFO₂ MW: 153.95 g/mol Bod tání: 216…222 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07363785 CAS číslo: 850593-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...