1-Bromo-2,4,5-trifluorobenzene 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,4,5-Trifluorobromobenzene
PC1580-25G
PC1580-500G
APOSPC1580-25GEA
1040
CZK
APOSPC1580-25G
APOSPC1580-500G
1-Bromo-2,4,5-trifluorobenzene 99%
1-Bromo-2,4,5-trifluorobenzene
For a study of lithiation under various conditions see: J. Chem. Soc, Perkin., 1,2729 (1995)
Vzorec:
C₆H₂BrF₃ MW: 210,98 g/mol Bod varu: 144 °C Bod tání: –19 °C Hustota: 1,802 g/cm³ (25 °C) Bod vzplanutí: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000306 CAS číslo: 327-52-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...