4-[(Trifluoromethyl)thio]benzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1572-25G
PC1572-100G
APOSPC1572-25GEA
7250
CZK
APOSPC1572-25G
APOSPC1572-100G
4-[(Trifluoromethyl)thio]benzonitrile 98%
4-[(Trifluoromethyl)thio]benzonitrile
Vzorec:
C₈H₄F₃NS MW: 203.19 g/mol Bod tání: 39…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039475 CAS číslo: 332-26-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...