Ethyl-2-bromo-2,3,3,3-tetrafluoropropionate 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC1567RM-5G
PC1567RM-1G
APOSPC1567RM-5GEA
6620
CZK
APOSPC1567RM-5G
APOSPC1567RM-1G
Ethyl-2-bromo-2,3,3,3-tetrafluoropropionate 97%
Ethyl-2-bromo-2,3,3,3-tetrafluoropropionate
Vzorec:
C₅H₅BrF₄O₂ Bod varu: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153721 CAS číslo: 10186-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...