5-Fluoro-2-nitroaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC1559-25G
PC1559-100G
PC1559-250G
APOSPC1559-25GEA
744
CZK
APOSPC1559-25G
APOSPC1559-100G
APOSPC1559-250G
5-Fluoro-2-nitroaniline
5-Fluoro-2-nitroaniline
Vzorec:
C₆H₅FN₂O₂ MW: 156,12 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00034065 CAS číslo: 2369-11-1 EINECS: 219-128-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...