Tisk…

1-Bromo-3,3,4,4,4-pentafluoro-2-butanone 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC1524-1G PC1524-5G
APOSPC1524-1GEA 3940 CZK
APOSPC1524-1G APOSPC1524-5G
1-Bromo-3,3,4,4,4-pentafluoro-2-butanone 98%
1-Bromo-3,3,4,4,4-pentafluoro-2-butanone
Vzorec: C₄H₂BrF₅O
MW: 240,96 g/mol
Bod varu: 93…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00041447
CAS číslo: 92737-01-4
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR