3-Fluoro-5-methylpyridine
Dodavatel: Apollo Scientific
PC1508-5G
PC1508-1G
PC1508-25G
APOSPC1508-5GEA
2170
CZK
APOSPC1508-5G
APOSPC1508-1G
APOSPC1508-25G
3-Fluoro-5-methylpyridine
3-Fluoro-5-methylpyridine
Vzorec:
C₆H₆FN MW: 111.12 Storage Temperature: Ambient |
MDL Number:
MFCD03095273 CAS číslo: 407-21-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...