2,4-Dichloro-3-fluorobenzonitrile 95+%
Dodavatel: Apollo Scientific
PC1505-10G
PC1505-5G
APOSPC1505-10GEA
7380
CZK
APOSPC1505-10G
APOSPC1505-5G
2,4-Dichloro-3-fluorobenzonitrile 95+%
2,4-Dichloro-3-fluorobenzonitrile
Vzorec:
C₇H₂Cl₂FN Storage Temperature: Ambient |
MDL Number:
MFCD06658268 CAS číslo: 161612-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...