2-Chloro-4,5-difluorobenzonitrile 98%
Dodavatel: Apollo Scientific
PC1504-25G
PC1504-5G
PC1504-50G
APOSPC1504-25GEA
5890
CZK
APOSPC1504-25G
APOSPC1504-5G
APOSPC1504-50G
2-Chloro-4,5-difluorobenzonitrile 98%
2-Chloro-4,5-difluorobenzonitrile
Vzorec:
C₇H₂ClF₂N MW: 173,55 g/mol Bod tání: 33…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD02871619 CAS číslo: 135748-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...