Tisk…

2-Bromo-4-(trifluoromethyl)anisole 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-4-methoxybenzotrifluoride, 2-Bromo-1-methoxy-4-(trifluoromethyl)benzene

PC1494-25G PC1494-5G
APOSPC1494-25GEA 2320 CZK
APOSPC1494-25G APOSPC1494-5G
2-Bromo-4-(trifluoromethyl)anisole 98%
2-Bromo-4-(trifluoromethyl)anisole
Vzorec: C₈H₆BrF₃O
MW: 255,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789249
CAS číslo: 402-10-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR