Tisk…

2-(Chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazole

Dodavatel: Apollo Scientific

PC1448-1G PC1448-5G
APOSPC1448-1GEA 4270 CZK
APOSPC1448-1G APOSPC1448-5G
2-(Chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazole
2-(Chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazole
Vzorec: C₁₀H₆ClF₃N₂O
Bod tání: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD06797380
CAS číslo: 753479-67-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR