5-Fluoro-2-pyridineboronic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Fluoropyridin-2-ylboronic acid
PC1428-1G
PC1428-250MG
APOSPC1428-1GEA
11180
CZK
APOSPC1428-1G
APOSPC1428-250MG
5-Fluoro-2-pyridineboronic acid 96%
5-Fluoro-2-pyridineboronic acid
Vzorec:
C₅H₅BFNO₂ Storage Temperature: Freezer |
MDL Number:
MFCD08705712 CAS číslo: 946002-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...