Tisk…

4-Bromo-4'-fluorobiphenyl 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Bromo-4'-fluoro-1,1'-biphenyl

PC1423J-1G
APOSPC1423J-1GEA 2690 CZK
APOSPC1423J-1G
4-Bromo-4'-fluorobiphenyl 99%
4-Bromo-4'-fluorobiphenyl
Vzorec: C₁₂H₈BrF
MW: 251,1 g/mol
Bod tání: 92…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00017954
CAS číslo: 398-21-0
EINECS: 206-909-0
UN: 3152
ADR: 9,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR