Tisk…

3-Bromo-3,3-difluoropropene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,3-Difluoro-3-bromopropene

PC1394-1G PC1394-25G PC1394-5G
APOSPC1394-1GEA 2280 CZK
APOSPC1394-1G APOSPC1394-25G APOSPC1394-5G
3-Bromo-3,3-difluoropropene 98%
3-Bromo-3,3-difluoropropene
Vzorec: C₃H₃BrF₂
MW: 156.96 g/mol
Bod varu: 42 °C
Hustota: 1.543 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042185
CAS číslo: 420-90-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR