1-Bromo-3,5-difluoro-2-iodobenzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC1392-100G
APOSPC1392-100GEA
3060
CZK
APOSPC1392-100G
1-Bromo-3,5-difluoro-2-iodobenzene 97%
1-Bromo-3,5-difluoro-2-iodobenzene
Vzorec:
C₆H₂BrF₂I MW: 318,89 g/mol Bod varu: 103…104 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00042183 CAS číslo: 175278-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...