Tisk…

(3-Bromo-2-fluorophenyl)acetonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Bromo-2-fluorophenylacetonitrile

PC1377-1G PC1377-25G PC1377-5G
APOSPC1377-1GEA 622 CZK
APOSPC1377-1G APOSPC1377-25G APOSPC1377-5G
(3-Bromo-2-fluorophenyl)acetonitrile 98%
(3-Bromo-2-fluorophenyl)acetonitrile
Vzorec: C₈H₅BrFN
MW: 214,04 g/mol
Bod tání: 25…26 °C
Storage Temperature: Ambient
MDL Number: MFCD09864662
CAS číslo: 874285-03-7
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR