Tisk…

1-Bromo-2-chloro-1,1,2-trifluoro-3-pentene 97%

Dodavatel: Apollo Scientific

PC1374KG-25G PC1374KG-5G
APOSPC1374KG-25GEA 34720 CZK
APOSPC1374KG-25G APOSPC1374KG-5G
1-Bromo-2-chloro-1,1,2-trifluoro-3-pentene 97%
1-Bromo-2-chloro-1,1,2-trifluoro-3-pentene
Vzorec: C₅H₅BrClF₃
Bod varu: 74…75 °C/93mmHg
Storage Temperature: Ambient
MDL Number: MFCD00153682
CAS číslo: 356-73-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR