2-Bromo-2-chloro-1,1,1-trifluoroethane 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Bromo-1-chloro-2,2,2-trifluoroethane
PC1372B-100G
PC1372B-25G
APOSPC1372B-100GEA
13030
CZK
APOSPC1372B-100G
APOSPC1372B-25G
2-Bromo-2-chloro-1,1,1-trifluoroethane 97%
2-Bromo-2-chloro-1,1,1-trifluoroethane
Vzorec:
C₂HBrClF₃ Bod varu: 50 °C Hustota: 1,872 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009602 CAS číslo: 151-67-7 UN: 3334 ADR: 9,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...