2-Bromo-4-fluoro-5-methylbenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC1372-1G
PC1372-250MG
APOSPC1372-1GEA
705
CZK
APOSPC1372-1G
APOSPC1372-250MG
2-Bromo-4-fluoro-5-methylbenzonitrile
2-Bromo-4-fluoro-5-methylbenzonitrile
Vzorec:
C₈H₅BrFN Storage Temperature: Ambient |
MDL Number:
MFCD07782070 CAS číslo: 916792-07-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...