1-Bromo-2-chloro-4-fluorobenzene 98+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-3-chloro-1-fluorobenzene, 2-Chloro-4-fluorobromobenzene, 4-Bromo-3-chlorofluorobenzene
PC1347-100G
PC1347-25G
PC1347-5G
PC1347-500G
APOSPC1347-100GEA
2240
CZK
APOSPC1347-100G
APOSPC1347-25G
APOSPC1347-5G
APOSPC1347-500G
1-Bromo-2-chloro-4-fluorobenzene 98+%
1-Bromo-2-chloro-4-fluorobenzene
Vzorec:
C₆H₃BrClF MW: 209,44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00142587 CAS číslo: 110407-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...